siRNA Details
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173546.2 | NP_775817.1 | 5, 6 | 1109 | ||
XM_005264938.2 | XP_005264995.1 | 5, 6 | 1595 | |||
XM_005264940.4 | XP_005264997.1 | 5, 6 | 1230 | |||
XM_006713015.2 | XP_006713078.1 | 5, 6 | 1625 | |||
XM_006713016.2 | XP_006713079.1 | 5, 6 | 1604 | |||
GenBank mRNA | AK294160.1 | - | 5, 6 | 958 | ||
AY129011.1 | - | 5, 6 | 1066 | |||
BC013110.2 | - | 1, 2 | 103 | |||
BC039083.1 | - | 5, 6 | 1059 | 5167173 |
