siRNA Details
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002909.3 | NP_001002909.1 | 8 | 1205 | ||
NM_001304939.1 | NP_001291868.1 | 7 | 1130 | |||
NM_001304940.1 | NP_001291869.1 | 10 | 1346 | |||
NM_001304941.1 | NP_001291870.1 | 9 | 1223 | |||
NM_001304942.1 | NP_001291871.1 | 7 | 1132 | |||
NM_001304943.1 | NP_001291872.1 | 5 | 1086 | |||
XM_011524557.1 | XP_011522859.1 | 10 | 1279 | |||
XM_011524558.1 | XP_011522860.1 | 11 | 1098 | |||
XM_011524559.2 | XP_011522861.1 | 12 | 1176 | |||
XM_011524561.2 | XP_011522863.1 | 8 | 937 | |||
XM_017024372.1 | XP_016879861.1 | 13 | 1221 | |||
XM_017024373.1 | XP_016879862.1 | 10 | 2056 | |||
XM_017024374.1 | XP_016879863.1 | 12 | 1139 | |||
XM_017024375.1 | XP_016879864.1 | 9 | 1234 | |||
XM_017024376.1 | XP_016879865.1 | 10 | 1094 | |||
XM_017024377.1 | XP_016879866.1 | 9 | 1071 | |||
XM_017024378.1 | XP_016879867.1 | 10 | 1252 | |||
XM_017024379.1 | XP_016879868.1 | 11 | 1134 | |||
XM_017024380.1 | XP_016879869.1 | 10 | 1072 | |||
GenBank mRNA | AB011125.2 | - | 7 | 2295 | ||
AK025600.1 | - | 8 | 1007 | |||
BC136629.1 | - | 9 | 1022 | 9020619 | ||
BC144323.1 | - | 9 | 1022 |
