siRNA Details
Gene Symbol: ZC3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015117.2 | NP_055932.2 | 2 | 397 | ||
XM_006716536.3 | XP_006716599.2 | 4 | 786 | |||
XM_011516943.2 | XP_011515245.1 | 2 | 512 | |||
XM_011516944.2 | XP_011515246.1 | 2 | 512 | |||
XM_017013248.1 | XP_016868737.1 | 3 | 1417 | |||
XM_017013249.1 | XP_016868738.1 | 3 | 959 | |||
GenBank mRNA | BC038670.1 | - | 2 | 380 | 5260824 | |
BM148116.1 | - | 2 | 400 | |||
D63484.1 | - | 2 | 357 |
