siRNA Details
Gene Symbol: IQCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287499.1 | NP_001274428.1 | 8, 9 | 834 | ||
NM_001287500.1 | NP_001274429.1 | 7, 8 | 786 | |||
NM_001287501.1 | NP_001274430.1 | 6, 7 | 740 | |||
NM_001287502.1 | NP_001274431.1 | 6, 7 | 740 | |||
NM_152558.4 | NP_689771.3 | 8, 9 | 834 | |||
XM_006715676.2 | XP_006715739.1 | 6, 7 | 588 | |||
XM_011515242.1 | XP_011513544.1 | 7, 8 | 693 | |||
XM_011515243.1 | XP_011513545.1 | 6, 7 | 715 | |||
XM_017011902.1 | XP_016867391.1 | 8, 9 | 896 | |||
XM_017011903.1 | XP_016867392.1 | 8, 9 | 774 | |||
GenBank mRNA | AB028946.2 | - | 6, 7 | 537 | ||
AK092404.1 | - | 8, 9 | 728 | |||
AK293372.1 | - | 7, 8 | 609 | |||
AK302053.1 | - | 6, 7 | 688 | |||
AK309646.1 | - | 8, 9 | 758 | |||
AL136792.1 | - | 7, 8 | 515 | |||
BC043150.1 | - | 6, 7 | 660 | 5297871 | ||
BC061518.1 | - | 7, 8 | 670 | |||
BC071858.1 | - | 8, 9 | 808 | 5478116 | ||
KJ534872.1 | - | 8, 9 | 624 | |||
KJ535081.1 | - | 6, 7 | 530 |
