siRNA Details
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 12 | 1500 | ||
NM_001281749.1 | NP_001268678.1 | 10 | 1232 | |||
NM_025164.4 | NP_079440.3 | 11 | 1376 | |||
XM_005271481.2 | XP_005271538.1 | 11 | 1414 | |||
XM_005271482.4 | XP_005271539.2 | 10 | 1270 | |||
XM_005271484.3 | XP_005271541.2 | 11 | 1414 | |||
XM_011542721.2 | XP_011541023.1 | 11 | 1376 | |||
XM_011542722.1 | XP_011541024.1 | 11 | 1414 | |||
XM_011542723.2 | XP_011541025.1 | 11 | 1257 | |||
XM_011542724.2 | XP_011541026.1 | 10 | 1111 | |||
XM_011542725.2 | XP_011541027.1 | 9 | 994 | |||
XM_011542726.1 | XP_011541028.1 | 11 | 1414 | |||
XM_017017424.1 | XP_016872913.1 | 10 | 1270 | |||
XM_017017425.1 | XP_016872914.1 | 11 | 1452 | |||
XM_017017426.1 | XP_016872915.1 | 9 | 1326 | |||
XM_017017427.1 | XP_016872916.1 | 7 | 683 | |||
GenBank mRNA | AB209327.1 | - | 4 | 386 | ||
BC128510.1 | - | 10 | 1094 | 40081639 | ||
BC128511.1 | - | 12 | 1356 | 40081640 |
