siRNA Details
Gene Symbol: TXLNGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045129.1 | 3, 4 | 510 | |||
GenBank mRNA | AF332224.1 | - | 3, 4 | 488 | ||
AI129169.1 | - | 1, 2 | 350 | |||
AK127387.1 | - | 3, 4 | 1757 | |||
BM263181.1 | - | 3, 4 | 510 |
