siRNA Details
Gene Symbol: ZNF396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322286.1 | NP_001309215.1 | 4 | 777 | ||
NM_001322290.1 | NP_001309219.1 | 5 | 1014 | |||
NM_145756.2 | NP_665699.1 | 4 | 777 | |||
XM_006722432.3 | XP_006722495.1 | 4 | 1505 | |||
XM_017025687.1 | XP_016881176.1 | 5 | 1557 | |||
XM_017025688.1 | XP_016881177.1 | 4 | 1392 | |||
XM_017025689.1 | XP_016881178.1 | 4 | 777 | |||
GenBank mRNA | AF533251.2 | - | 4 | 777 | ||
AK055775.1 | - | 4 | 777 | |||
BC130288.1 | - | 4 | 764 | 40146319 | ||
BC136314.1 | - | 4 | 764 | 9020301 |
