siRNA Details
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077491.1 | NP_001070959.1 | 4 | 426 | ||
NM_001077492.1 | NP_001070960.1 | 3 | 358 | |||
NM_012427.4 | NP_036559.1 | 3 | 561 | |||
XM_011526702.1 | XP_011525004.1 | 3 | 300 | |||
XM_011526703.2 | XP_011525005.1 | 4 | 380 | |||
GenBank mRNA | AF168768.2 | - | 3 | 377 | ||
AF435980.1 | - | 3 | 532 | |||
AF435981.1 | - | 3 | 329 | |||
AK223421.1 | - | 3 | 509 | |||
AY279380.1 | - | 3 | 335 | |||
AY279381.1 | - | 4 | 403 | |||
AY359010.1 | - | 3 | 561 | |||
AY461805.1 | - | 2 | 219 | |||
BC008036.2 | - | 3 | 492 | 2989806 | ||
BT006867.1 | - | 2 | 208 |
