siRNA Details
Gene Symbol: TAS2R39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176881.2 | NP_795362.2 | 1 | 612 | ||
GenBank mRNA | BC100947.2 | - | 1 | 612 | ||
BC100948.2 | - | 1 | 612 | |||
BC114952.1 | - | 1 | 612 |
