siRNA Details
Gene Symbol: C20orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009984.2 | NP_001009984.1 | 23 | 2066 | ||
XM_005260684.3 | XP_005260741.1 | 23 | 2060 | |||
XM_005260687.4 | XP_005260744.1 | 4 | 902 | |||
XM_011529208.2 | XP_011527510.1 | 4 | 403 | |||
XM_017027783.1 | XP_016883272.1 | 20 | 1687 | |||
GenBank mRNA | AK023919.1 | - | 5 | 728 | ||
AK226100.1 | - | 18 | 1490 | |||
AK303375.1 | - | 14 | 1435 | |||
AL110249.1 | - | 14 | 1031 | |||
BC122529.1 | - | 14 | 1284 | 40012183 |
