siRNA Details
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 4 | 632 | ||
NM_001271869.1 | NP_001258798.1 | 4 | 629 | |||
NR_073500.1 | 2 | 327 | ||||
NR_073501.1 | 5 | 703 | ||||
NR_073502.1 | 3 | 582 | ||||
NR_073503.1 | 3 | 579 | ||||
GenBank mRNA | AB097042.1 | - | 3 | 370 | ||
AB097043.1 | - | 3 | 370 | |||
AB097044.1 | - | 3 | 361 | |||
AB097045.1 | - | 3 | 361 | |||
AB097046.1 | - | 4 | 420 | |||
AJ295987.1 | - | 3 | 366 | |||
AK308787.1 | - | 2 | 114 | |||
AL050028.1 | - | 3 | 366 | |||
BC023988.1 | - | 3 | 387 | 3825796 | ||
BC065538.1 | - | 3 | 367 | 6053254 | ||
DA010145.1 | - | 4 | 424 | |||
DA438597.1 | - | 5 | 490 |
