siRNA Details
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 13 | 1950 | ||
GenBank mRNA | AF135016.1 | - | 12 | 1314 | ||
BC011180.1 | - | 7 | 727 | |||
BM676246.1 | - | 1 | 183 | |||
BM715684.1 | - | 7 | 690 | |||
CB218994.1 | - | 2 | 143 |
