siRNA Details
Gene Symbol: ZNF776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173632.3 | NP_775903.3 | 3 | 797 | ||
GenBank mRNA | AK127764.1 | - | 2 | 443 | ||
AK299680.1 | - | 3 | 797 | |||
BC136752.1 | - | 2 | 439 | 9020742 | ||
BC136753.1 | - | 2 | 439 | 9020743 | ||
CR936802.1 | - | 3 | 751 |
