siRNA Details
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076678.2 | NP_001070146.1 | 4 | 934 | ||
NM_175910.6 | NP_787106.4 | 2 | 711 | |||
GenBank mRNA | AB209086.1 | - | 2 | 679 | ||
AK127476.1 | - | 1 | 319 | |||
AK128494.1 | - | 2 | 697 | |||
BC112346.1 | - | 1 | 530 | |||
BX648842.1 | - | 4 | 924 |
