siRNA Details
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512262.2 | XP_011510564.1 | 1, 2 | 397 | ||
XM_017005474.1 | XP_016860963.1 | 1, 2 | 397 | |||
XM_017005475.1 | XP_016860964.1 | 1, 2 | 397 | |||
XM_017005476.1 | XP_016860965.1 | 2, 3 | 412 | |||
XM_017005477.1 | XP_016860966.1 | 2, 3 | 2007 | |||
XM_017005478.1 | XP_016860967.1 | 2, 3 | 138 | |||
GenBank mRNA | BC041437.1 | - | 2, 3 | 130 |
