siRNA Details
Gene Symbol: ZNF621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098414.2 | NP_001091884.1 | 3, 4 | 367 | ||
NM_001287245.1 | NP_001274174.1 | 3, 4 | 603 | |||
NM_198484.4 | NP_940886.1 | 3, 4 | 603 | |||
GenBank mRNA | AK074366.1 | - | 3, 4 | 528 | ||
AK127181.1 | - | 3, 4 | 367 | |||
BC101619.1 | - | 3, 4 | 241 | 8069125 | ||
BC113415.1 | - | 3, 4 | 241 | 8322467 | ||
CR749268.1 | - | 3, 4 | 468 | |||
DA282973.1 | - | 3, 4 | 367 | |||
DB480668.2 | - | 3, 4 | 593 |
