siRNA Details
Gene Symbol: SMLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195597.1 | NP_001182526.1 | 1 | 228 | ||
GenBank mRNA | AK091900.1 | - | 1 | 227 | ||
BC101422.1 | - | 1 | 227 | |||
BC101423.1 | - | 1 | 227 | |||
BC101424.1 | - | 1 | 227 | |||
BC101425.1 | - | 1 | 227 | |||
BC113026.1 | - | 1 | 227 | |||
BC113027.1 | - | 1 | 228 | |||
BX095531.1 | - | 1 | 166 |
