siRNA Details
Gene Symbol: COA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040431.2 | NP_001035521.1 | 1, 2 | 260 | ||
GenBank mRNA | AF070665.1 | - | 1, 2 | 235 | ||
AK225611.1 | - | 1, 2 | 239 | |||
AK290863.1 | - | 1, 2 | 226 | |||
AL524532.3 | - | 1, 2 | 260 | |||
AU122748.1 | - | 1, 2 | 226 | |||
BC002698.2 | - | 1, 2 | 221 | 3609536 |
