siRNA Details
Gene Symbol: KRTAP6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181605.3 | NP_853636.3 | 1 | 258 | ||
GenBank mRNA | AB096951.1 | - | 1 | 258 | ||
BC101121.2 | - | 1 | 232 | |||
BC101122.1 | - | 1 | 232 | |||
BC118547.1 | - | 1 | 232 | |||
BC118624.1 | - | 1 | 232 |
