siRNA Details
Gene Symbol: C2orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105519.2 | NP_001098989.1 | 4 | 572 | ||
NM_001322426.1 | NP_001309355.1 | 5 | 608 | |||
XM_011532810.2 | XP_011531112.1 | 6 | 775 | |||
XM_011532811.2 | XP_011531113.1 | 6 | 2271 | |||
XM_017003962.1 | XP_016859451.1 | 7 | 703 | |||
XM_017003963.1 | XP_016859452.1 | 5 | 647 | |||
XM_017003964.1 | XP_016859453.1 | 6 | 679 | |||
XM_017003965.1 | XP_016859454.1 | 6 | 626 | |||
XM_017003966.1 | XP_016859455.1 | 7 | 852 | |||
GenBank mRNA | AA725022.1 | - | 1 | 179 | ||
AA758869.1 | - | 1 | 181 | |||
AK091483.1 | - | 3 | 2184 | |||
BX114846.1 | - | 5 | 581 |
