siRNA Details
Gene Symbol: CLPSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010886.3 | NP_001010886.1 | 2, 3 | 284 | ||
XM_017010820.1 | XP_016866309.1 | 1, 2 | 363 | |||
GenBank mRNA | AI018488.1 | - | 1, 2 | 286 | ||
BC133046.1 | - | 2, 3 | 284 | 40147068 | ||
BC137392.1 | - | 2, 3 | 284 | 9021390 | ||
BX106742.1 | - | 2, 3 | 294 | |||
DQ823638.1 | - | 2, 3 | 295 |
