siRNA Details
Gene Symbol: RGAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024455.3 | NP_001019626.1 | 1 | 1323 | ||
GenBank mRNA | AB082532.1 | - | 1 | 1323 | ||
AL359579.2 | - | 1 | 367 | |||
BC151224.1 | - | 1 | 1344 | 9007468 |
