siRNA Details
Gene Symbol: C16orf96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145011.1 | NP_001138483.1 | 5 | 1772 | ||
XM_005255298.3 | XP_005255355.1 | 6 | 2299 | |||
XM_005255301.3 | XP_005255358.1 | 5 | 2359 | |||
XM_011522475.2 | XP_011520777.1 | 7 | 2282 |
