siRNA Details
Gene Symbol: ZNF850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193552.1 | NP_001180481.1 | 5 | 540 | ||
NM_001267779.1 | NP_001254708.1 | 4 | 444 | |||
GenBank mRNA | BC052603.1 | - | 5 | 540 | ||
BC136266.1 | - | 5 | 478 | |||
BC143637.1 | - | 4 | 382 |
