siRNA Details
Gene Symbol: NCCRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001414.1 | NP_001001414.1 | 5, 6 | 706 | ||
GenBank mRNA | AK123941.1 | - | 5, 6 | 706 | ||
BC067874.1 | - | 5, 6 | 706 | 30348184 | ||
BC092493.1 | - | 5, 6 | 709 | 30337042 |
