siRNA Details
Gene Symbol: ZNF677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317998.1 | NP_001304927.1 | 5 | 872 | ||
NM_182609.3 | NP_872415.1 | 5 | 690 | |||
XM_005258862.4 | XP_005258919.1 | 5 | 906 | |||
XM_011526912.2 | XP_011525214.1 | 5 | 818 | |||
GenBank mRNA | AK308120.1 | - | 5 | 652 | ||
BC050038.1 | - | 5 | 645 | 5272569 | ||
BX648243.1 | - | 5 | 661 |
