siRNA Details
Gene Symbol: GPR148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207364.2 | NP_997247.2 | 1 | 783 | ||
GenBank mRNA | AY255532.1 | - | 1 | 515 | ||
AY280965.1 | - | 2 | 599 | |||
AY569570.1 | - | 1 | 703 | |||
BC105013.1 | - | 1 | 783 | 8144016 | ||
BC105041.1 | - | 1 | 783 | 8144044 |
