siRNA Details
Gene Symbol: FAM159A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042693.2 | NP_001036158.1 | 2 | 441 | ||
NR_135156.1 | 3 | 534 | ||||
NR_135157.1 | 2 | 441 | ||||
XM_011541377.2 | XP_011539679.1 | 2 | 1710 | |||
XM_011541378.2 | XP_011539680.1 | 2 | 1711 | |||
GenBank mRNA | AK128236.1 | - | 2 | 441 | ||
AY358621.1 | - | 2 | 215 | |||
BC041608.1 | - | 3 | 484 | |||
BG192125.1 | - | 1 | 113 |
