siRNA Details
Gene Symbol: FAM131C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182623.2 | NP_872429.2 | 7 | 770 | ||
GenBank mRNA | AK094085.1 | - | 7 | 659 | ||
AL529663.3 | - | 6 | 653 | |||
BC016848.1 | - | 7 | 764 | 3913592 | ||
BC037394.1 | - | 7 | 738 | 5198517 | ||
BG751253.1 | - | 3 | 308 |
