siRNA Details
Gene Symbol: ITGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002204.3 | NP_002195.1 | 4, 5 | 1126 | ||
XM_005257308.2 | XP_005257365.1 | 4, 5 | 1029 | |||
GenBank mRNA | AB209658.1 | - | 4, 5 | 1026 | ||
AK289961.1 | - | 4, 5 | 752 | |||
AK303384.1 | - | 5, 6 | 680 | |||
BC136636.1 | - | 4, 5 | 726 | 9020626 | ||
BC144328.1 | - | 4, 5 | 726 | 9052848 | ||
BC150190.1 | - | 4, 5 | 969 | 40148521 | ||
D01038.1 | - | 4, 5 | 558 | |||
M59911.1 | - | 4, 5 | 727 |
