siRNA Details
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198516.2 | NP_940918.2 | 3, 4 | 1012 | ||
XM_006718224.2 | XP_006718287.1 | 3, 4 | 1043 | |||
XM_006718225.2 | XP_006718288.1 | 3, 4 | 1043 | |||
XM_011520068.2 | XP_011518370.1 | 3, 4 | 588 | |||
XM_011520069.2 | XP_011518371.1 | 3, 4 | 1043 | |||
XM_011520070.2 | XP_011518372.1 | 3, 4 | 1043 | |||
XM_011520071.2 | XP_011518373.1 | 3, 4 | 1043 | |||
GenBank mRNA | AB078147.1 | - | 3, 4 | 591 | ||
AJ626724.1 | - | 3, 4 | 591 | |||
AK055111.1 | - | 3, 4 | 994 | |||
BC037341.2 | - | 3, 4 | 1001 | 5265475 | ||
BC060864.1 | - | 3, 4 | 1003 | 30341070 |
