siRNA Details
Gene Symbol: C3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198562.2 | NP_940964.1 | 1 | 567 | ||
GenBank mRNA | AK125642.1 | - | 1 | 511 | ||
BC023586.2 | - | 1 | 1401 | 4650297 | ||
BC053629.1 | - | 1 | 567 | 6204001 | ||
BC061697.1 | - | 1 | 557 | 6737728 |
