siRNA Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242800.1 | NP_001229729.1 | 4 | 316 | ||
NM_001242801.1 | NP_001229730.1 | 4 | 494 | |||
NM_001242802.1 | NP_001229731.1 | 4 | 584 | |||
NM_206894.3 | NP_996777.2 | 4 | 276 | |||
XM_005258903.4 | XP_005258960.1 | 4 | 643 | |||
XM_011526950.2 | XP_011525252.1 | 4 | 724 | |||
GenBank mRNA | AK297474.1 | - | 4 | 285 | ||
BC057245.1 | - | 4 | 278 | 6712908 | ||
BM473790.1 | - | 2 | 116 | |||
DA819130.1 | - | 4 | 274 | |||
DC307556.1 | - | 4 | 492 | |||
DC308617.1 | - | 4 | 318 | |||
DC352882.1 | - | 4 | 540 | |||
DC377558.1 | - | 4 | 508 | |||
EL735861.1 | - | 2 | 155 |
