siRNA Details
Gene Symbol: FAM150A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207413.3 | NP_997296.1 | 4 | 550 | ||
GenBank mRNA | AY358213.1 | - | 4 | 553 | ||
BC130641.1 | - | 4 | 497 | 40146672 | ||
BC130643.1 | - | 4 | 497 | 40146674 |
