siRNA Details
Gene Symbol: OR4D10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004705.1 | NP_001004705.1 | 1 | 74 | ||
GenBank mRNA | BC136893.1 | - | 1 | 131 | 9020884 | |
BC136894.1 | - | 1 | 131 | 9020885 |
