siRNA Details
Gene Symbol: MUC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010909.3 | NP_001010909.2 | 2 | 1738 | ||
NM_001322370.1 | NP_001309299.1 | 1804 | ||||
NM_001322371.1 | NP_001309300.1 | 1911 | ||||
NR_130720.1 | 2 | 1880 | ||||
GenBank mRNA | AB242595.1 | - | 2 | 1648 | ||
AK056612.1 | - | 1 | 233 | |||
AK298642.1 | - | 2 | 1726 | |||
AK299471.1 | - | 1911 | ||||
AK301645.1 | - | 1 | 1446 | |||
AK316363.1 | - | 2 | 1728 | |||
AY358415.1 | - | 1815 | ||||
BC105735.1 | - | 2 | 1673 | 40026480 | ||
BC105736.1 | - | 2 | 1815 | |||
BC105737.1 | - | 2 | 1673 | 40026476 | ||
BC107478.1 | - | 3 | 1223 | 40026469 | ||
HM596784.1 | - | 2 | 1667 |
