siRNA Details
Gene Symbol: C11orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100388.1 | NP_001093858.1 | 4 | 357 | ||
NM_207430.2 | NP_997313.2 | 5 | 438 | |||
GenBank mRNA | AI829856.1 | - | 3 | 359 | ||
AK128145.1 | - | 5 | 438 | |||
BC039505.2 | - | 4 | 342 |
