siRNA Details
Gene Symbol: C17orf97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013672.4 | NP_001013694.4 | 2 | 570 | ||
GenBank mRNA | AK128660.1 | - | 570 | |||
BC057385.1 | - | 2 | 521 | |||
BC070116.1 | - | 2 | 496 | |||
BC141806.1 | - | 2 | 569 | 40147362 | ||
KR780251.1 | - | 2 | 570 |
Gene Symbol: LOC105371430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136407.1 | 2 | 58 | |||
GenBank mRNA | BI029000.1 | - | 2 | 58 |
