siRNA Details
Gene Symbol: FAM177B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324080.1 | NP_001311009.1 | 5 | 625 | ||
NM_207468.2 | NP_997351.2 | 6 | 727 | |||
NR_136691.1 | 6 | 1231 | ||||
XM_017001279.1 | XP_016856768.1 | 5 | 1169 | |||
XM_017001280.1 | XP_016856769.1 | 6 | 683 | |||
XM_017001281.1 | XP_016856770.1 | 6 | 1252 | |||
XM_017001282.1 | XP_016856771.1 | 6 | 708 | |||
GenBank mRNA | AK125494.1 | - | 6 | 1231 |
