siRNA Details
Gene Symbol: LDLRAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013693.2 | NP_001013715.2 | 4 | 926 | ||
XM_005245873.4 | XP_005245930.1 | 4 | 982 | |||
GenBank mRNA | AK127275.1 | - | 4 | 926 | ||
BC146860.1 | - | 4 | 828 | 9056788 |
