siRNA Details
Gene Symbol: OR6C65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005518.1 | NP_001005518.1 | 1 | 417 | ||
GenBank mRNA | BC136897.1 | - | 1 | 516 | 9020888 | |
BC136899.1 | - | 1 | 516 | 9020890 |
