siRNA Details
Gene Symbol: CUEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271875.1 | NP_001258804.1 | 5, 6 | 1194 | ||
NM_001292025.1 | NP_001278954.1 | 5, 6 | 1499 | |||
XM_006721900.2 | XP_006721963.1 | 5, 6 | 1809 | |||
XM_011524812.2 | XP_011523114.1 | 5, 6 | 582 | |||
XM_017024641.1 | XP_016880130.1 | 6, 7 | 1211 | |||
GenBank mRNA | AK000746.1 | - | 5, 6 | 1499 | ||
BC056882.1 | - | 5, 6 | 1194 | 6150503 |
