siRNA Details
Gene Symbol: OOEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080507.2 | NP_001073976.1 | 2, 3 | 378 | ||
XM_017010890.1 | XP_016866379.1 | 2, 3 | 357 | |||
GenBank mRNA | BX093821.1 | - | 2, 3 | 368 | ||
EU290647.1 | - | 2, 3 | 378 |
