siRNA Details
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517881.2 | XP_011516183.1 | 21 | 3497 | ||
XM_011517883.2 | XP_011516185.1 | 20 | 3443 | |||
XM_011517885.2 | XP_011516187.1 | 15 | 2588 | |||
XM_017014728.1 | XP_016870217.1 | 15 | 2585 |
