siRNA Details
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716749.3 | XP_006716812.1 | 21 | 5168 | ||
XM_011517836.2 | XP_011516138.1 | 21 | 5304 | |||
XM_011517837.2 | XP_011516139.1 | 21 | 5301 | |||
GenBank mRNA | AK058117.1 | - | 1 | 366 | ||
BC024252.1 | - | 1 | 393 | |||
BC047488.1 | - | 1 | 389 |
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517881.2 | XP_011516183.1 | 21 | 5044 | ||
XM_011517883.2 | XP_011516185.1 | 20 | 4990 | |||
XM_011517885.2 | XP_011516187.1 | 15 | 4135 | |||
XM_017014728.1 | XP_016870217.1 | 15 | 4132 |
