siRNA Details
Gene Symbol: NDUFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004545.3 | NP_004536.2 | 1, 2 | 176 | ||
GenBank mRNA | AF050638.1 | - | 1, 2 | 30 | ||
BC009691.1 | - | 1, 2 | 36 | |||
BC104672.1 | - | 1, 2 | 96 | |||
BC126232.1 | - | 1, 2 | 173 | 8991948 | ||
BC126234.1 | - | 1, 2 | 173 | 8991950 | ||
BM052981.1 | - | 1, 2 | 176 | |||
BX476462.1 | - | 1, 2 | 75 |
