siRNA Details
Gene Symbol: PLEKHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197026.1 | NP_001183955.1 | 14 | 1866 | ||
XM_011515596.2 | XP_011513898.1 | 13 | 1382 | |||
XM_017012746.1 | XP_016868235.1 | 14 | 1873 | |||
XM_017012747.1 | XP_016868236.1 | 14 | 1906 | |||
GenBank mRNA | AF308300.1 | - | 14 | 1543 | ||
AK294857.1 | - | 14 | 1815 |
Gene Symbol: PLEKHA8P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037144.1 | 3 | 1653 | |||
GenBank mRNA | AF103731.1 | - | 3 | 1653 | ||
AF380162.1 | - | 1 | 1482 | |||
AK024218.1 | - | 1 | 628 | |||
BC028914.1 | - | 1 | 1494 | |||
BC063575.1 | - | 3 | 1534 | 3881751 | ||
BU741550.1 | - | 1 | 231 |

More Information
Other Species Targeted

Gene Ontology Categories:
