siRNA Details
Gene Symbol: PLEKHA8P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037144.1 | 3 | 1697 | |||
GenBank mRNA | AF103731.1 | - | 3 | 1697 | ||
AF380162.1 | - | 1 | 1526 | |||
AK024218.1 | - | 1 | 672 | |||
BC028914.1 | - | 1 | 1538 | |||
BC063575.1 | - | 3 | 1578 | 3881751 | ||
BU741550.1 | - | 1 | 187 |
