siRNA Details
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 5 | 612 | ||
GenBank mRNA | AF113537.1 | - | 5 | 481 | ||
AF123767.1 | - | 5 | 481 | |||
AF133422.1 | - | 5 | 465 | |||
AK098398.1 | - | 1020 | ||||
AK294383.1 | - | 4 | 395 | |||
AK295206.1 | - | 4 | 295 | |||
AK297196.1 | - | 5 | 418 | |||
AK312359.1 | - | 5 | 499 | |||
AK315949.1 | - | 3 | 227 | |||
AK315952.1 | - | 4 | 309 | |||
BC002619.2 | - | 5 | 495 | 3160345 | ||
BC018873.2 | - | 5 | 488 | |||
CR457142.1 | - | 4 | 350 |
