siRNA Details
Gene Symbol: SELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016275.3 | NP_057359.2 | 4 | 531 | ||
GenBank mRNA | AF131856.1 | - | 4 | 486 | ||
AF195141.1 | - | 4 | 489 | |||
AK075006.1 | - | 4 | 504 | |||
AK300315.1 | - | 4 | 519 | |||
AY358095.1 | - | 4 | 495 | |||
AY358096.1 | - | 4 | 499 | |||
BC006012.1 | - | 4 | 522 | 4333083 | ||
BC008411.1 | - | 4 | 489 | 4132578 | ||
BC009556.1 | - | 4 | 463 | 3899430 | ||
BC009611.1 | - | 4 | 483 | 4093231 | ||
BC026350.1 | - | 4 | 511 | 4796883 | ||
BC036738.1 | - | 4 | 482 | 5540243 | ||
BC071699.1 | - | 4 | 474 | 5548565 | ||
CR457154.1 | - | 3 | 273 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001040396, AK146778, BC043068, AK032838, BC132453, and others.
Gene Ontology Categories:
